4-(4-Brom-3-(trifluormethyl)phenylsulfonyl)morpholin

Lieferant: COMBI-BLOCKS

HC-6471-5G HC-6471-1G
COBBHC-6471-5GEA 983 EUR
COBBHC-6471-5G COBBHC-6471-1G
4-(4-Brom-3-(trifluormethyl)phenylsulfonyl)morpholin
4-(4-Brom-3-(trifluormethyl)phenylsulfonyl)morpholin
Formel: C₁₁H₁₁BrF₃NO₃S
MDL: MFCD09972160
CAS-Nummer: 1020253-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR