4-(3-Brom-4-methylphenylsulfonyl)morpholin

Lieferant: COMBI-BLOCKS

Synonyme: 4-((3-Bromo-4-methylphenyl)sulphonyl)morpholine

OT-0168-5G OT-0168-1G
COBBOT-0168-5GEA 656 EUR
COBBOT-0168-5G COBBOT-0168-1G
4-(3-Brom-4-methylphenylsulfonyl)morpholin
4-(3-Brom-4-methylphenylsulfonyl)morpholin
Formel: C₁₁H₁₄BrNO₃S
Molecular Weight: 320,2028 g/mol
MDL: MFCD07363839
CAS-Nummer: 850429-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR