4-(4-Bromphenylsulfonyl)morpholin

Lieferant: COMBI-BLOCKS

Synonyme: 4-((4-Bromophenyl)sulphonyl)morpholine

OT-0089-5G OT-0089-25G OT-0089-1G
COBBOT-0089-5GEA 141 EUR
COBBOT-0089-5G COBBOT-0089-25G COBBOT-0089-1G
4-(4-Bromphenylsulfonyl)morpholin
4-(4-Bromphenylsulfonyl)morpholin
Formel: C₁₀H₁₂BrNO₃S
Molecular Weight: 306.1800 g/mol
MDL: MFCD00194739
CAS-Nummer: 834-67-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR