3-(Morpholinosulfonyl)anilin
Lieferant: COMBI-BLOCKS
OT-0136-5G
OT-0136-1G
COBBOT-0136-5GEA
460
EUR
COBBOT-0136-5G
COBBOT-0136-1G
3-(Morpholinosulfonyl)anilin
3-(Morpholinosulfonyl)anilin
Formel:
C₁₀H₁₄N₂O₃S Molecular Weight: 242.300 g/mol |
MDL:
MFCD02090909 CAS-Nummer: 22184-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...