4-(2-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((2-Chlorobenzyl)oxy)-3,5-dimethylphenyl)boronic acid
BB-8112-1G
BB-8112-250M
COBBBB-8112-1GEA
353
EUR
COBBBB-8112-1G
COBBBB-8112-250M
4-(2-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
4-(2-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
Formel:
C₁₅H₁₆BClO₃ Molecular Weight: 290,5500 g/mol |
MDL:
MFCD06411361 CAS-Nummer: 849052-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...