4-(3-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)boronic acid
BB-8113-1G
BB-8113-250M
COBBBB-8113-1GEA
451
EUR
COBBBB-8113-1G
COBBBB-8113-250M
4-(3-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
4-(3-Chlorbenzyloxy)-3,5-dimethylphenylboronsäure
Formel:
C₁₅H₁₆BClO₃ Molecular Weight: 290,5500 g/mol |
MDL:
MFCD06411362 CAS-Nummer: 849062-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...