2-Benzyloxy-5-brom-1,3-dimethylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Benzyloxy)-5-bromo-1,3-dimethylbenzene

OT-0760-5G OT-0760-1G
COBBOT-0760-5GEA 303 EUR
COBBOT-0760-5G COBBOT-0760-1G
2-Benzyloxy-5-brom-1,3-dimethylbenzol
2-Benzyloxy-5-brom-1,3-dimethylbenzol
Formel: C₁₅H₁₅BrO
MDL: MFCD12913972
CAS-Nummer: 168196-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR