(3-Aminopropyl)diethoxymethylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL371890-50MLEA 55.5 EUR
SIAL371890-50ML SIAL371890-250ML
(3-Aminopropyl)diethoxymethylsilan, Sigma-Aldrich®
(3-Aminopropyl)diethoxymethylsilan
Formel: C₈H₂₁NO₂Si
Molecular Weight: 191,35 g/mol
MDL: MFCD00039785
CAS-Nummer: 3179-76-8
EINECS: 221-660-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR