(3-Aminopropyl)diethoxymethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Diethoxy(methyl)silyl)propan-1-amine

SILS00775.25 SILS00775.100 SILS00775.100
SILNSILS00775.25EA 0 EUR
SILNSILS00775.25 CHMPSILS00775.100 SILNSILS00775.100
(3-Aminopropyl)diethoxymethylsilan
(3-Aminopropyl)diethoxymethylsilan
Formel: C₈H₂₁NO₂Si
Molecular Weight: 191.35 g/mol
MDL: MFCD00039785
CAS-Nummer: 3179-76-8
EINECS: 221-660-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR