3-(2-Aminoethylamino)propylmethyldimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
3-(2-Aminoethylamino)propyldimethoxymethylsilane
SILS00600.25
SILS00600.100
SILS00600.500
SILNSILS00600.25EA
0
EUR
SILNSILS00600.25
SILNSILS00600.100
SILNSILS00600.500
3-(2-Aminoethylamino)propylmethyldimethoxysilan
3-(2-Aminoethylamino)propylmethyldimethoxysilan
Formel:
C₈H₂₂N₂O₂Si Molecular Weight: 206,36 g/mol |
MDL:
MFCD00054776 CAS-Nummer: 3069-29-2 EINECS: 221-336-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...