(3-Aminopropyl)dimethylmethoxysilan

Lieferant: CHEMPUR SILANE

SILS00760.5 SILS00760.10
SILNSILS00760.5EA 0 EUR
SILNSILS00760.5 SILNSILS00760.10
(3-Aminopropyl)dimethylmethoxysilan
(3-Aminopropyl)dimethylmethoxysilan
Formel: C₆H₁₇NOSi
MDL: MFCD01861689
CAS-Nummer: 31024-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR