(Trimethylstannyl)dimethylvinylsilan
Lieferant: FLUOROCHEM
|
Gefahr
|
009695-1G
009695-5G
FLUO009695-1GEA
0
EUR
FLUO009695-1G
FLUO009695-5G
(Trimethylstannyl)dimethylvinylsilan
(Trimethylstannyl)dimethylvinylsilan
Formel:
C₇H₁₈SiSn |
MDL:
MFCD00800632 CAS-Nummer: 214279-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...