(Trimethylstannyl)dimethylvinylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20418.1 SILS20418.5
SILNSILS20418.1EA 0 EUR
SILNSILS20418.1 SILNSILS20418.5
(Trimethylstannyl)dimethylvinylsilan
(Trimethylstannyl)dimethylvinylsilan
Formel: C₇H₁₈SiSn
MDL: MFCD00800632
CAS-Nummer: 214279-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR