(Trimethylstannyl)dimethylphenylsilan
Lieferant: FLUOROCHEM
|
Gefahr
|
009694-1G
009694-5G
FLUO009694-1GEA
0
EUR
FLUO009694-1G
FLUO009694-5G
(Trimethylstannyl)dimethylphenylsilan
(Trimethylstannyl)dimethylphenylsilan
Formel:
C₁₁H₂₀SiSn |
MDL:
MFCD00800633 CAS-Nummer: 94397-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...