(Trimethylstannyl)dimethylphenylsilan
Lieferant: CHEMPUR SILANE
|
Gefahr
|
SILS20412.1
SILS20412.5
SILNSILS20412.1EA
0
EUR
SILNSILS20412.1
SILNSILS20412.5
(Trimethylstannyl)dimethylphenylsilan
(Trimethylstannyl)dimethylphenylsilan
Formel:
C₁₁H₂₀SiSn |
MDL:
MFCD00800633 CAS-Nummer: 94397-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...