(Trimethylstannyl)butyldimethylsilan

Lieferant: FLUOROCHEM
Gefahr

009693-1G 009693-5G
FLUO009693-1GEA 0 EUR
FLUO009693-1G FLUO009693-5G
(Trimethylstannyl)butyldimethylsilan
(Trimethylstannyl)butyldimethylsilan
Formel: C₉H₂₄SiSn
MDL: MFCD03093828
CAS-Nummer: 103731-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR