(Trimethylstannyl)diphenylmethylsilan
Lieferant: FLUOROCHEM
|
Gefahr
|
009696-1G
009696-5G
FLUO009696-1GEA
0
EUR
FLUO009696-1G
FLUO009696-5G
(Trimethylstannyl)diphenylmethylsilan
(Trimethylstannyl)diphenylmethylsilan
Formel:
C₁₆H₂₂SiSn |
MDL:
MFCD00800634 CAS-Nummer: 149013-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...