(Trimethylstannyl)diphenylmethylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20415.1 SILS20415.5
SILNSILS20415.1EA 0 EUR
SILNSILS20415.1 SILNSILS20415.5
(Trimethylstannyl)diphenylmethylsilan
(Trimethylstannyl)diphenylmethylsilan
Formel: C₁₆H₂₂SiSn
MDL: MFCD00800634
CAS-Nummer: 149013-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR