(Trimethylstannyl)triphenylsilan

Lieferant: FLUOROCHEM
Gefahr

009697-1G 009697-5G
FLUO009697-1GEA 0 EUR
FLUO009697-1G FLUO009697-5G
(Trimethylstannyl)triphenylsilan
(Trimethylstannyl)triphenylsilan
Formel: C₂₁H₂₄SiSn
MDL: MFCD03093829
CAS-Nummer: 18748-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR