(Trimethylstannyl)triphenylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20420.1 SILS20420.5
SILNSILS20420.1EA 0 EUR
SILNSILS20420.1 SILNSILS20420.5
(Trimethylstannyl)triphenylsilan
(Trimethylstannyl)triphenylsilan
Formel: C₂₁H₂₄SiSn
MDL: MFCD03093829
CAS-Nummer: 18748-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR