(Triphenylstannyl)dimethylvinylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20720.1 SILS20720.5 009700-1G
SILNSILS20720.1EA 0 EUR
SILNSILS20720.1 SILNSILS20720.5 FLUO009700-1G
(Triphenylstannyl)dimethylvinylsilan
(Triphenylstannyl)dimethylvinylsilan
Formel: C₂₂H₂₄SiSn
MDL: MFCD03093832
CAS-Nummer: 210362-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR