(Triphenylstannyl)triphenylsilan

Lieferant: FLUOROCHEM
Gefahr

009702-1G 009702-5G
FLUO009702-1GEA 0 EUR
FLUO009702-1G FLUO009702-5G
(Triphenylstannyl)triphenylsilan
(Triphenylstannyl)triphenylsilan
Formel: C₃₆H₃₀SiSn
MDL: MFCD03093834
CAS-Nummer: 6928-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR