(Triphenylstannyl)triphenylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20723.1 SILS20723.5
SILNSILS20723.1EA 0 EUR
SILNSILS20723.1 SILNSILS20723.5
(Triphenylstannyl)triphenylsilan
(Triphenylstannyl)triphenylsilan
Formel: C₃₆H₃₀SiSn
MDL: MFCD03093834
CAS-Nummer: 6928-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR