(±)-Boc-α-phosphonoglycintrimethylester 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

B25421.03 B25421.14 B25421.06
B25421.03EA 55.6 EUR
B25421.03 B25421.14 B25421.06
(±)-Boc-α-phosphonoglycintrimethylester 95%
(±)-Boc-α-phosphonoglycintrimethylester

Purity: 95% CAS No.: 89524-98-1 Molecular Formula: C10H20NO7P Formula Weight: 297.24 Melting Point: 67-71° MDL No.: MFCD02178893

Formel: C₁₀H₂₀NO₇P
Molecular Weight: 297.24 g/mol
Schmelzpunkt: 67…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02178893
CAS-Nummer: 89524-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR