2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril

Lieferant: BIONET RESEARCH

3G-029.500 3G-029.1 3G-029.5 3G-029.10
BIOT3G-029.500EA 0 EUR
BIOT3G-029.500 BIOT3G-029.1 BIOT3G-029.5 BIOT3G-029.10
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril
Formel: C₁₄H₆Cl₃F₃N₂
MDL: MFCD00172210
CAS-Nummer: 338407-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR