2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-phenylacetonitril
Lieferant: BIONET RESEARCH
3G-002.500
3G-002.1
3G-002.5
3G-002.10
BIOT3G-002.500EA
0
EUR
BIOT3G-002.500
BIOT3G-002.1
BIOT3G-002.5
BIOT3G-002.10
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-phenylacetonitril
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-phenylacetonitril
Formel:
C₁₄H₈ClF₃N₂ |
MDL:
MFCD00172206 CAS-Nummer: 213994-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...