2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitril
Lieferant: BIONET RESEARCH
3G-038.500
3G-038.1
3G-038.5
3G-038.10
BIOT3G-038.500EA
0
EUR
BIOT3G-038.500
BIOT3G-038.1
BIOT3G-038.5
BIOT3G-038.10
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitril
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitril
Formel:
C₁₄H₇ClF₃N₃O₂ |
MDL:
MFCD00172215 CAS-Nummer: 213994-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...