2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitril

Lieferant: BIONET RESEARCH

3G-032.500 3G-032.1 3G-032.5 3G-032.10
BIOT3G-032.500EA 0 EUR
BIOT3G-032.500 BIOT3G-032.1 BIOT3G-032.5 BIOT3G-032.10
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitril
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitril
Formel: C₁₅H₁₀ClF₃N₂O
MDL: MFCD00172212
CAS-Nummer: 338407-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR