N(α)-Boc-L-tryptophan
Lieferant: NOVABIOCHEM
Synonyme: Boc-Trp-OH
8.53038.0100 8.53038.0025
04-12-0006-100EA 266 EUR
04-12-0006-100 04-12-0006-25
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304.3 g/mol
MDL: MFCD00065595
CAS-Nummer: 13139-14-5
EINECS: 236-072-7