α-Methylzimtsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methyl-3-phenyl-2-propenoic acid
M-Clarity™ Qualitätsstufe = MQ100
M35606-100G
M35606-25G
SIALM35606-100GEA
314
EUR
SIALM35606-100G
SIALM35606-25G
α-Methylzimtsäure, Sigma-Aldrich®
α-Methylzimtsäure
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol |
MDL:
MFCD00002652 CAS-Nummer: 1199-77-5 EINECS: 214-847-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...