2-Brom-4,5-dimethoxyzimtsäure

Lieferant: MENAI

IP12.50 IP12.100
MENAIP12.50EA 0 EUR
MENAIP12.50 MENAIP12.100
2-Brom-4,5-dimethoxyzimtsäure
2-Brom-4,5-dimethoxyzimtsäure
Formel: C₁₀H₉BrO₃
Molecular Weight: 287,12 g/mol
MDL: MFCD00017614
CAS-Nummer: 151539-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR