3-(Trifluormethoxy)zimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(3-(Trifluoromethoxy)phenyl)acrylic acid
009359-25G
009359-5G
FLUO009359-25GEA
134
EUR
FLUO009359-25G
FLUO009359-5G
3-(Trifluormethoxy)zimtsäure
3-(Trifluormethoxy)zimtsäure
Formel:
C₁₀H₇F₃O₃ Molecular Weight: 232.16 g/mol |
MDL:
MFCD00066337 CAS-Nummer: 168833-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...