1-(3-Brom-5-nitrobenzoyl)piperidin
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-5-nitrophenyl)(piperidin-1-yl)methanone
CA-4379-5G
CA-4379-1G
COBBCA-4379-5GEA
0
EUR
COBBCA-4379-5G
COBBCA-4379-1G
1-(3-Brom-5-nitrobenzoyl)piperidin
1-(3-Brom-5-nitrobenzoyl)piperidin
Formel:
C₁₂H₁₃BrN₂O₃ |
MDL:
MFCD09475858 CAS-Nummer: 941294-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...