1-(3-Brom-4-fluorbenzoyl)piperidin

Lieferant: COMBI-BLOCKS

CA-4613-5G CA-4613-1G CA-4613-25G
COBBCA-4613-5GEA 3100 EUR
COBBCA-4613-5G COBBCA-4613-1G COBBCA-4613-25G
1-(3-Brom-4-fluorbenzoyl)piperidin
1-(3-Brom-4-fluorbenzoyl)piperidin
Formel: C₁₂H₁₃BrFNO
MDL: MFCD10699648
CAS-Nummer: 1033201-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR