1-(4-Brom-2-fluorbenzoyl)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-2-fluorobenzoyl)piperidine

CA-4759-5G CA-4759-1G
COBBCA-4759-5GEA 451 EUR
COBBCA-4759-5G COBBCA-4759-1G
1-(4-Brom-2-fluorbenzoyl)piperidin
1-(4-Brom-2-fluorbenzoyl)piperidin
Formel: C₁₂H₁₃BrFNO
MDL: MFCD09938596
CAS-Nummer: 1017035-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR