4-(4-Methoxybenzoyl)piperidin

Lieferant: CHEMPUR

60-0115.500 60-0115.1
CHMP60-0115.500EA 0 EUR
CHMP60-0115.500 CHMP60-0115.1
4-(4-Methoxybenzoyl)piperidin
4-(4-Methoxybenzoyl)piperidin
Formel: C₁₃H₁₇NO₂
MDL: MFCD00835819
CAS-Nummer: 76362-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR