1-(4-Fluorbenzoyl)pyrrolidin

Lieferant: COMBI-BLOCKS

CA-4542-1G CA-4542-25G CA-4542-5G
COBBCA-4542-1GEA 0 EUR
COBBCA-4542-1G COBBCA-4542-25G COBBCA-4542-5G
1-(4-Fluorbenzoyl)pyrrolidin
1-(4-Fluorbenzoyl)pyrrolidin
Formel: C₁₁H₁₂FNO
MDL: MFCD00465428
CAS-Nummer: 349644-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR