4-(4-Fluorbenzoyl)piperidin

Lieferant: CHEMPUR

60-0103.1 60-0103.5
CHMP60-0103.1EA 0 EUR
CHMP60-0103.1 CHMP60-0103.5
4-(4-Fluorbenzoyl)piperidin
4-(4-Fluorbenzoyl)piperidin
Formel: C₁₂H₁₄FNO
MDL: MFCD03411595
CAS-Nummer: 56346-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR