4-(4-Fluorbenzoyl)piperidin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: (4-Fluorophenyl)(piperidin-4-yl)methanone hydrochloride

001709-5G 001709-25G
FLUO001709-5GEA 55.4 EUR
FLUO001709-5G FLUO001709-25G
4-(4-Fluorbenzoyl)piperidin Hydrochlorid
4-(4-Fluorbenzoyl)piperidin Hydrochlorid
Formel: C₁₂H₁₅ClFNO
Molecular Weight: 243,71 g/mol
MDL: MFCD00044912
CAS-Nummer: 25519-78-2
EINECS: 247-070-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR