(S)-3-(Aminomethyl)-1-Boc-piperidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (S)-3-(aminomethyl)piperidine-1-carboxylate, (S)-1-BOC-3-(Aminomethyl)piperidine

AM-1764-5G AM-1764-1G
COBBAM-1764-5GEA 201 EUR
COBBAM-1764-5G COBBAM-1764-1G
(S)-3-(Aminomethyl)-1-Boc-piperidin
(S)-3-(Aminomethyl)-1-Boc-piperidin
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,31 g/mol
MDL: MFCD03839877
CAS-Nummer: 140645-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR