1-Boc-4-methylpiperidin

Lieferant: CHEMPUR

Synonyme: tert-Butyl 4-methylpiperidine-1-carboxylate

60-0002.1 60-0002.5
CHMP60-0002.1EA 0 EUR
CHMP60-0002.1 CHMP60-0002.5
1-Boc-4-methylpiperidin
1-Boc-4-methylpiperidin
Formel: C₁₁H₂₁NO₂
Molecular Weight: 199,2899 g/mol
MDL: MFCD04114947
CAS-Nummer: 123387-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR