1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

4R-1332.500 4R-1332.1 4R-1332.5 4R-1332.10
BIOT4R-1332.500EA 0 EUR
BIOT4R-1332.500 BIOT4R-1332.1 BIOT4R-1332.5 BIOT4R-1332.10
1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄BrNO₄S
MDL: MFCD02331440
CAS-Nummer: 203519-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR