1-Brom-3-fluor-5-pyrrolidinobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(3-Bromo-5-fluorophenyl)pyrrolidine

AN-1502-1G AN-1502-5G
COBBAN-1502-1GEA 188 EUR
COBBAN-1502-1G COBBAN-1502-5G
1-Brom-3-fluor-5-pyrrolidinobenzol
1-Brom-3-fluor-5-pyrrolidinobenzol
Formel: C₁₀H₁₁BrFN
Molecular Weight: 244,1035 g/mol
MDL: MFCD13195637
CAS-Nummer: 1199773-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR