1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-(piperidin-1-ylsulphonyl)benzotrifluoride

HC-6310-5G HC-6310-1G
COBBHC-6310-5GEA 451 EUR
COBBHC-6310-5G COBBHC-6310-1G
1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin
1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin
Formel: C₁₂H₁₃BrF₃NO₂S
MDL: MFCD09801013
CAS-Nummer: 951884-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR