1-((4-Brom-3-(trifluormethyl)phenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

HC-6474-5G HC-6474-1G
COBBHC-6474-5GEA 1180 EUR
COBBHC-6474-5G COBBHC-6474-1G
1-((4-Brom-3-(trifluormethyl)phenyl)sulfonyl)piperidin
1-((4-Brom-3-(trifluormethyl)phenyl)sulfonyl)piperidin
Formel: C₁₂H₁₃BrF₃NO₂S
Molecular Weight: 372,2000 g/mol
MDL: MFCD11053849
CAS-Nummer: 1065074-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR