1-((2-Bromphenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

HC-6158-5G HC-6158-1G
COBBHC-6158-5GEA 595 EUR
COBBHC-6158-5G COBBHC-6158-1G
1-((2-Bromphenyl)sulfonyl)piperidin
1-((2-Bromphenyl)sulfonyl)piperidin
Formel: C₁₁H₁₄BrNO₂S
MDL: MFCD09800969
CAS-Nummer: 951883-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR