1-((3-Brom-4-methylphenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

OT-0167-1G OT-0167-5G
COBBOT-0167-1GEA 149 EUR
COBBOT-0167-1G COBBOT-0167-5G
1-((3-Brom-4-methylphenyl)sulfonyl)piperidin
1-((3-Brom-4-methylphenyl)sulfonyl)piperidin
Formel: C₁₂H₁₆BrNO₂S
Molecular Weight: 318,2299 g/mol
MDL: MFCD07363838
CAS-Nummer: 850429-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR