1-((3-Brom-5-methylphenyl)sulfonyl)piperidin

Lieferant: COMBI-BLOCKS

HC-6460-1G HC-6460-5G
COBBHC-6460-1GEA 149 EUR
COBBHC-6460-1G COBBHC-6460-5G
1-((3-Brom-5-methylphenyl)sulfonyl)piperidin
1-((3-Brom-5-methylphenyl)sulfonyl)piperidin
Formel: C₁₂H₁₆BrNO₂S
MDL: MFCD09972151
CAS-Nummer: 1020252-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR