1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid

Lieferant: BIONET RESEARCH

1R-1300.500 1R-1300.1 1R-1300.5 1R-1300.10
BIOT1R-1300.500EA 0 EUR
BIOT1R-1300.500 BIOT1R-1300.1 BIOT1R-1300.5 BIOT1R-1300.10
1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
Formel: C₁₂H₁₆ClN₃O₃S
MDL: MFCD00754333
CAS-Nummer: 332400-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR