1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
1-((4-Chlorophenyl)sulphonyl)piperidine-4-carboxylic acid
4R-1137.500
4R-1137.1
4R-1137.5
4R-1137.10
BIOT4R-1137.500EA
0
EUR
BIOT4R-1137.500
BIOT4R-1137.1
BIOT4R-1137.5
BIOT4R-1137.10
1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Chlorphenyl)sulfonyl)-4-piperidincarbonsäure
Formel:
C₁₂H₁₄ClNO₄S Molecular Weight: 303,7619 g/mol |
MDL:
MFCD00723388 CAS-Nummer: 314744-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...